Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lfng | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008494.3 | NP_032520.1 | 8 | 1844 | ||
GenBank mRNA | AK004642.1 | - | 8 | 1845 | ||
BQ571136.1 | - | 1 | 653 |