Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pparg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011146.3 | NP_035276.2 | 3 | 384 | ||
GenBank mRNA | AB644275.1 | - | 4 | 390 | ||
AY208183.1 | - | 3 | 384 | |||
AY208184.1 | - | 3 | 384 | |||
AY243585.1 | - | 3 | 384 | |||
BC021798.1 | - | 3 | 384 | 4191539 | ||
CJ043379.1 | - | 5 | 459 | |||
CN701591.1 | - | 4 | 394 | |||
CT010340.1 | - | 3 | 339 | |||
EF062476.1 | - | 3 | 375 | |||
GQ868647.1 | - | 2 | 142 | |||
GQ868648.1 | - | 2 | 122 | |||
GQ868649.1 | - | 2 | 142 | |||
GQ868650.1 | - | 2 | 122 | |||
U01664.1 | - | 4 | 416 | |||
U01841.1 | - | 4 | 739 | |||
U09138.1 | - | 3 | 378 | |||
U10374.1 | - | 2 | 249 |