siRNA Details
Gene Symbol: Sox18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009236.2 | NP_033262.2 | 2 | 1492 | ||
GenBank mRNA | AK144281.1 | - | 2 | 1493 | ||
AV167860.1 | - | 1 | 223 | |||
BC006612.1 | - | 2 | 1492 | 3967084 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
