Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022312.3 | NP_071707.2 | 20, 21 | 4248 | ||
GenBank mRNA | AK039390.1 | - | 20, 21 | 4249 | ||
AK078229.1 | - | 13, 14 | 1926 | |||
BC132392.1 | - | 20, 21 | 3882 | 40130669 | ||
BC138043.1 | - | 20, 21 | 3882 | 8861062 |