siRNA Details
Gene Symbol: Tnr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022312.3 | NP_071707.2 | 12 | 3019 | ||
GenBank mRNA | AK039390.1 | - | 12 | 3020 | ||
AK078229.1 | - | 5 | 697 | |||
BC132392.1 | - | 12 | 2653 | 40130669 | ||
BC138043.1 | - | 12 | 2653 | 8861062 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
