Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdpd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201352.2 | NP_958740.2 | 5, 6 | 1107 | ||
GenBank mRNA | AK141189.1 | - | 5, 6 | 1140 | ||
AK154771.1 | - | 5, 6 | 1244 | |||
BC024955.1 | - | 4, 5 | 393 | 4953174 | ||
BC026428.1 | - | 1, 2 | 40 | 4224574 | ||
BC082585.1 | - | 5, 6 | 1107 | 30608279 |