Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pask | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080850.2 | NP_543126.2 | 10, 11 | 3211 | ||
GenBank mRNA | AK028435.1 | - | 10, 11 | 3213 | ||
AK083528.1 | - | 10, 11 | 3182 | |||
AK083590.1 | - | 10, 11 | 3177 | |||
AK129063.1 | - | 10, 11 | 3101 | |||
AK172031.1 | - | 10, 11 | 3159 | |||
BC150821.1 | - | 9, 10 | 2923 | 9087732 |