Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fastkd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146084.1 | NP_001139556.1 | 3 | 2219 | ||
NM_198176.2 | NP_937819.2 | 3 | 2256 | |||
GenBank mRNA | AK143929.1 | - | 3 | 2195 | ||
AK220420.1 | - | 1 | 1820 | |||
BC048956.1 | - | 1 | 1258 | |||
BC052898.1 | - | 3 | 2176 | 30059761 |