Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Utp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023054.1 | NP_075541.1 | 1 | 1354 | ||
GenBank mRNA | AF155362.2 | - | 1 | 1349 | ||
AF271213.1 | - | 1 | 1354 | |||
AK010316.1 | - | 1 | 1371 | |||
AK145637.1 | - | 1 | 1367 | |||
BC022994.1 | - | 1 | 1359 | 5371012 |