Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fibin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026271.1 | NP_080547.1 | 1 | 1335 | ||
GenBank mRNA | AK003790.1 | - | 1 | 1335 | ||
AK011548.1 | - | 1 | 1506 | |||
AK159648.1 | - | 1 | 1468 | |||
BC027250.1 | - | 1 | 1324 | 3603588 |