Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026785.2 | NP_081061.1 | 3, 4 | 268 | ||
GenBank mRNA | AK003722.1 | - | 3, 4 | 269 | ||
AK136186.1 | - | 3, 4 | 269 | |||
AK160740.1 | - | 3, 4 | 300 | |||
BC147032.1 | - | 3, 4 | 249 | 9056036 | ||
BC147033.1 | - | 3, 4 | 249 | 9056038 | ||
BC171983.1 | - | 3, 4 | 242 |