Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC17A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC017952.1 | - | 10 | 1196 | 4557795 | |
BX107915.1 | - | 1 | 73 | |||
U90545.1 | - | 10 | 1436 |