Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnk10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029911.4 | NP_084187.2 | 7 | 1542 | ||
GenBank mRNA | AK031904.1 | - | 7 | 1473 | ||
AK052327.1 | - | 1 | 405 | |||
AK082153.1 | - | 7 | 1543 | |||
BC132487.1 | - | 7 | 1484 | 40130764 | ||
BC137869.1 | - | 7 | 1484 | 8860888 | ||
DQ185134.1 | - | 7 | 1439 |