Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023876.3 | NP_076365.2 | 8 | 1020 | ||
GenBank mRNA | AJ276004.1 | - | 8 | 1019 | ||
AK034040.1 | - | 8 | 1025 | |||
AK034529.1 | - | 8 | 1026 | |||
AK037104.1 | - | 8 | 1044 | |||
AK039447.1 | - | 8 | 1032 | |||
AK039758.1 | - | 8 | 1021 | |||
AK079069.1 | - | 8 | 1035 | |||
AK168964.1 | - | 8 | 1084 |