Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VAMP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006634.2 | NP_006625.1 | 2, 3 | 227 | ||
GenBank mRNA | AF054825.1 | - | 2, 3 | 201 | ||
AF077197.1 | - | 1, 2 | 154 | |||
AF151025.1 | - | 2, 3 | 185 | |||
AK292660.1 | - | 2, 3 | 225 | |||
BC017891.1 | - | 2, 3 | 227 | 4617039 | ||
CR542215.1 | - | 1, 2 | 144 |