Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCR9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256369.1 | NP_001243298.1 | 4 | 501 | ||
NM_006641.3 | NP_006632.2 | 2 | 366 | |||
NM_031200.2 | NP_112477.1 | 3 | 415 | |||
XM_011533312.2 | XP_011531614.1 | 3 | 1128 | |||
GenBank mRNA | AF145439.1 | - | 3 | 392 | ||
AF145440.1 | - | 2 | 311 | |||
AJ132337.1 | - | 3 | 294 | |||
AK312824.1 | - | 4 | 456 | |||
AK313366.1 | - | 3 | 369 | |||
BC069678.1 | - | 3 | 376 | 7262627 | ||
BC095516.1 | - | 1 | 223 | |||
DB103756.1 | - | 3 | 415 | |||
DB132476.1 | - | 2 | 361 |