Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270364.1 | NP_001257293.1 | 3 | 405 | ||
NM_001270365.1 | NP_001257294.1 | 3 | 507 | |||
NM_012190.3 | NP_036322.2 | 3 | 507 | |||
NR_072979.1 | 3 | 507 | ||||
XM_006713481.2 | XP_006713544.1 | 3 | 246 | |||
XM_011512355.1 | XP_011510657.1 | 3 | 239 | |||
XM_017005613.1 | XP_016861102.1 | 3 | 630 | |||
XM_017005614.1 | XP_016861103.1 | 3 | 246 | |||
GenBank mRNA | AB209299.1 | - | 3 | 314 | ||
AF052732.1 | - | 3 | 267 | |||
AK222535.1 | - | 3 | 289 | |||
AK222694.1 | - | 3 | 289 | |||
AK294392.1 | - | 3 | 291 | |||
AK307699.1 | - | 4 | 418 | |||
AK308112.1 | - | 3 | 246 | |||
BC027241.1 | - | 3 | 280 | 4484301 | ||
BI829048.1 | - | 3 | 347 | |||
CR749807.1 | - | 3 | 405 | |||
DC342268.1 | - | 3 | 505 | |||
KM111188.1 | - | 3 | 316 | |||
KU178540.1 | - | 2 | 157 | |||
KU178541.1 | - | 3 | 341 | |||
KU178542.1 | - | 2 | 157 |