Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHRS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282987.1 | NP_001269916.1 | 4 | 483 | ||
NM_001282988.1 | NP_001269917.1 | 4 | 452 | |||
NM_001282990.1 | NP_001269919.1 | 4 | 452 | |||
NM_021004.3 | NP_066284.2 | 5 | 554 | |||
XM_006720005.3 | XP_006720068.1 | 5 | 554 | |||
GenBank mRNA | AB045131.1 | - | 5 | 579 | ||
AF044127.1 | - | 5 | 525 | |||
AF064256.1 | - | 5 | 528 | |||
AK001870.1 | - | 5 | 520 | |||
AK314448.1 | - | 5 | 519 | |||
AY358638.1 | - | 5 | 535 | |||
AY616182.1 | - | 4 | 483 | |||
BC003019.1 | - | 5 | 502 | 2822884 | ||
DQ325464.1 | - | 4 | 419 | |||
DQ338571.1 | - | 3 | 281 | |||
DQ344810.1 | - | 5 | 533 |
Gene Symbol: DHRS4L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193635.1 | NP_001180564.1 | 6 | 588 | ||
NM_001193636.1 | NP_001180565.1 | 5 | 831 | |||
NM_198083.3 | NP_932349.2 | 5 | 630 | |||
GenBank mRNA | AK021587.1 | - | 1 | 320 | ||
BC000663.2 | - | 5 | 538 | 3349226 | ||
DQ088987.1 | - | 3 | 129 | |||
DQ088988.1 | - | 3 | 129 | |||
GU564234.1 | - | 6 | 496 | |||
GU564235.1 | - | 5 | 598 |