Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTNL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_197975.2 | NP_932079.1 | 2 | 406 | ||
GenBank mRNA | AB020625.1 | - | 2 | 380 | ||
AK025267.1 | - | 2 | 384 | |||
AY358385.1 | - | 2 | 355 | |||
BC100800.1 | - | 2 | 292 | 40000697 | ||
BC100801.1 | - | 2 | 292 | 40000698 | ||
BC100802.1 | - | 2 | 292 | 40000699 | ||
DA431389.1 | - | 2 | 406 |