Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRSF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032102.3 | NP_115285.1 | 1 | 885 | ||
NR_103726.1 | 1 | 885 | ||||
XM_017017142.1 | XP_016872631.1 | 1 | 847 | |||
GenBank mRNA | AF031166.1 | - | 1 | 505 | ||
AK023379.1 | - | 1 | 771 | |||
AK312514.1 | - | 1 | 774 | |||
BC057783.1 | - | 2 | 575 | 5285882 | ||
CV805885.1 | - | 1 | 268 |