Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MORF4L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001265603.1 | NP_001252532.1 | 2 | 304 | ||
NM_001265604.1 | NP_001252533.1 | 4 | 372 | |||
NM_001265605.1 | NP_001252534.1 | 4 | 272 | |||
NM_006791.3 | NP_006782.1 | 4 | 419 | |||
NM_206839.2 | NP_996670.1 | 5 | 536 | |||
XM_017021872.1 | XP_016877361.1 | 4 | 384 | |||
GenBank mRNA | AF070664.1 | - | 4 | 226 | ||
AF100615.1 | - | 4 | 313 | |||
AF109188.1 | - | 4 | 208 | |||
AF161546.1 | - | 4 | 205 | |||
AF167173.1 | - | 4 | 221 | |||
AF218011.1 | - | 4 | 324 | |||
AK095823.1 | - | 5 | 821 | |||
AK098485.1 | - | 4 | 339 | |||
AK296650.1 | - | 5 | 427 | |||
AK300789.1 | - | 4 | 356 | |||
AK310181.1 | - | 3 | 157 | |||
AK310592.1 | - | 4 | 218 | |||
AK310702.1 | - | 4 | 227 | |||
AL137697.1 | - | 4 | 213 | |||
AY148481.1 | - | 5 | 430 | |||
BC002936.2 | - | 4 | 300 | 3955120 | ||
BC022845.2 | - | 4 | 300 | 2988803 | ||
BC067826.1 | - | 4 | 195 | 4824022 | ||
BC141840.1 | - | 4 | 227 | 8860303 | ||
BG771854.1 | - | 4 | 429 | |||
BX382045.2 | - | 4 | 216 | |||
CX165647.1 | - | 2 | 55 | |||
DB455429.1 | - | 4 | 272 |