Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MORF4L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001265603.1 | NP_001252532.1 | 8 | 850 | ||
NM_001265604.1 | NP_001252533.1 | 10 | 918 | |||
NM_001265605.1 | NP_001252534.1 | 10 | 818 | |||
NM_006791.3 | NP_006782.1 | 10 | 965 | |||
NM_206839.2 | NP_996670.1 | 11 | 1082 | |||
XM_017021872.1 | XP_016877361.1 | 10 | 930 | |||
GenBank mRNA | AF070664.1 | - | 10 | 772 | ||
AF100615.1 | - | 10 | 859 | |||
AF109188.1 | - | 10 | 754 | |||
AF131847.1 | - | 6 | 488 | |||
AF161546.1 | - | 10 | 751 | |||
AF167173.1 | - | 10 | 767 | |||
AF218011.1 | - | 10 | 870 | |||
AK095823.1 | - | 11 | 1367 | |||
AK098485.1 | - | 10 | 885 | |||
AK296650.1 | - | 11 | 973 | |||
AK300789.1 | - | 10 | 902 | |||
AL137697.1 | - | 10 | 758 | |||
AY148481.1 | - | 11 | 976 | |||
BC002936.2 | - | 10 | 846 | 3955120 | ||
BC022845.2 | - | 10 | 846 | 2988803 | ||
BC067826.1 | - | 10 | 741 | 4824022 | ||
BC121092.1 | - | 6 | 468 | 40119977 | ||
BC121093.1 | - | 6 | 468 | 40119978 | ||
BC141840.1 | - | 10 | 773 | 8860303 | ||
BX382045.2 | - | 10 | 762 | |||
CX165647.1 | - | 8 | 601 |