Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MORF4L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001265603.1 | NP_001252532.1 | 6 | 588 | ||
NM_001265604.1 | NP_001252533.1 | 8 | 656 | |||
NM_001265605.1 | NP_001252534.1 | 8 | 556 | |||
NM_006791.3 | NP_006782.1 | 8 | 703 | |||
NM_206839.2 | NP_996670.1 | 9 | 820 | |||
XM_017021872.1 | XP_016877361.1 | 8 | 668 | |||
GenBank mRNA | AF070664.1 | - | 8 | 510 | ||
AF100615.1 | - | 8 | 597 | |||
AF109188.1 | - | 8 | 492 | |||
AF131847.1 | - | 4 | 226 | |||
AF161546.1 | - | 8 | 489 | |||
AF167173.1 | - | 8 | 505 | |||
AF218011.1 | - | 8 | 608 | |||
AK095823.1 | - | 9 | 1105 | |||
AK098485.1 | - | 8 | 623 | |||
AK296650.1 | - | 9 | 711 | |||
AK300789.1 | - | 8 | 640 | |||
AK310181.1 | - | 6 | 910 | |||
AK310702.1 | - | 7 | 430 | |||
AL137697.1 | - | 8 | 497 | |||
AY148481.1 | - | 9 | 714 | |||
BC002936.2 | - | 8 | 584 | 3955120 | ||
BC022845.2 | - | 8 | 584 | 2988803 | ||
BC067826.1 | - | 8 | 479 | 4824022 | ||
BC121092.1 | - | 4 | 206 | 40119977 | ||
BC121093.1 | - | 4 | 206 | 40119978 | ||
BC141840.1 | - | 8 | 511 | 8860303 | ||
BX382045.2 | - | 8 | 500 | |||
CX165647.1 | - | 6 | 339 |