Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282444.1 | NP_001269373.1 | 6 | 1465 | ||
NM_001282445.1 | NP_001269374.1 | 5 | 1143 | |||
NM_006795.3 | NP_006786.2 | 4 | 1322 | |||
XM_011544739.1 | XP_011543041.1 | 5 | 1140 | |||
GenBank mRNA | AF001434.1 | - | 4 | 1273 | ||
AF099011.1 | - | 1264 | ||||
AK097842.1 | - | 6 | 1430 | |||
AK098723.1 | - | 3 | 593 | |||
AK124613.1 | - | 3 | 2240 | |||
AK126581.1 | - | 2 | 900 | |||
AK297654.1 | - | 5 | 1001 | |||
AK299096.1 | - | 5 | 1054 | |||
AK300326.1 | - | 5 | 1103 | |||
AK302930.1 | - | 5 | 1095 | |||
AK312316.1 | - | 4 | 1175 | |||
AY007161.1 | - | 4 | 1069 | |||
BC104799.1 | - | 4 | 1214 | 8143802 | ||
BC104825.1 | - | 4 | 1214 | 8143828 |