Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOMM34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006809.4 | NP_006800.2 | 6 | 852 | ||
XM_005260254.2 | XP_005260311.1 | 6 | 785 | |||
XM_011528501.1 | XP_011526803.1 | 6 | 671 | |||
XM_017027600.1 | XP_016883089.1 | 5 | 751 | |||
GenBank mRNA | AB085681.1 | - | 6 | 777 | ||
AK000594.1 | - | 6 | 771 | |||
AK129538.1 | - | 5 | 353 | |||
AK222952.1 | - | 6 | 759 | |||
AK302128.1 | - | 6 | 681 | |||
AK307655.1 | - | 6 | 793 | |||
BC001763.1 | - | 6 | 765 | 3544124 | ||
BC007423.2 | - | 6 | 765 | 2960090 | ||
BC014907.1 | - | 6 | 742 | 3922502 | ||
BT020008.1 | - | 6 | 712 | |||
BT020009.1 | - | 6 | 712 | |||
CR456765.1 | - | 6 | 712 | |||
U58970.1 | - | 6 | 743 |