Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLLT11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006818.3 | NP_006809.1 | 2 | 1771 | ||
GenBank mRNA | AK056089.1 | - | 2 | 1766 | ||
AK057755.1 | - | 3 | 1004 | |||
BC006471.2 | - | 1 | 893 | 2823316 | ||
BC007079.1 | - | 2 | 1035 | |||
BC008445.1 | - | 2 | 1035 | 4103485 | ||
BC009624.1 | - | 2 | 1051 | 3878959 | ||
BC021703.1 | - | 2 | 1109 | 4108071 | ||
BC022448.1 | - | 2 | 2043 | 4815214 | ||
BC032996.1 | - | 2 | 1068 | |||
U16954.1 | - | 2 | 1243 |