Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SF3B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006842.2 | NP_006833.2 | 9, 10 | 1012 | ||
XM_005273726.3 | XP_005273783.1 | 9, 10 | 1582 | |||
XM_011544740.2 | XP_011543042.1 | 9, 10 | 1582 | |||
XM_017017144.1 | XP_016872633.1 | 9, 10 | 1579 | |||
GenBank mRNA | AK290850.1 | - | 9, 10 | 996 | ||
AK300016.1 | - | 3, 4 | 286 | |||
BC000401.2 | - | 9, 10 | 971 | |||
BC007610.1 | - | 2, 3 | 195 | 3351485 | ||
BC053577.1 | - | 9, 10 | 979 | |||
BX537771.1 | - | 7, 8 | 685 | |||
U41371.1 | - | 9, 10 | 951 |