Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006859.3 | NP_006850.2 | 7, 8 | 828 | ||
NM_194451.2 | NP_919433.1 | 7, 8 | 828 | |||
XM_017007666.1 | XP_016863155.1 | 4, 5 | 521 | |||
GenBank mRNA | AK026289.1 | - | 7, 8 | 796 | ||
AK292238.1 | - | 7, 8 | 828 | |||
BC023635.2 | - | 7, 8 | 773 | 4841470 |