Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OGFR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007346.2 | NP_031372.2 | 4 | 388 | ||
GenBank mRNA | AF172449.1 | - | 4 | 386 | ||
AF172450.1 | - | 4 | 386 | |||
AF172451.1 | - | 4 | 386 | |||
AF172452.1 | - | 4 | 386 | |||
AF172453.1 | - | 386 | ||||
AK022234.1 | - | 4 | 378 | |||
AK024485.1 | - | 2 | 2474 | |||
AK314696.1 | - | 4 | 380 | |||
BC008768.2 | - | 4 | 363 | 3050950 | ||
BC014137.2 | - | 4 | 363 | 4643904 | ||
BC032666.1 | - | 4 | 343 | 5552531 | ||
BM551600.1 | - | 4 | 388 |