Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABHD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007011.7 | NP_008942.3 | 14 | 1959 | ||
NM_152924.4 | NP_690888.1 | 10 | 1561 | |||
XM_005254832.1 | XP_005254889.1 | 13 | 1597 | |||
XM_005254833.1 | XP_005254890.1 | 10 | 1205 | |||
XM_011521165.1 | XP_011519467.1 | 11 | 1410 | |||
GenBank mRNA | AF546700.1 | - | 10 | 1293 | ||
AK222841.1 | - | 10 | 1233 | |||
AK223083.1 | - | 10 | 1231 | |||
AK290340.1 | - | 10 | 1233 | |||
AK296827.1 | - | 8 | 816 | |||
BC019248.1 | - | 14 | 1689 | 4826878 | ||
BC090052.1 | - | 10 | 1295 | 30388486 | ||
X12433.1 | - | 10 | 1216 |