Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SERPINA1 SIRNA Assays ›
Gene Symbol:
Serpina1
Gene Name:
serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 1
Gene Aliases:
AAT, Pi, Spi1
Chromosome Location:
Chr.6: 127610241 - 127632265 on Build Rnor_6.0
UniGene ID:
Rn.1419
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
s217870
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Serpina1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_022519.2
NP_071964.2
3
765
XM_006240456.1
XP_006240518.1
2
622
GenBank mRNA
BC078824.1
-
3
765
7131535
D00675.1
-
2
568
FQ209479.1
-
1
79
FQ209494.1
-
1
79
FQ209517.1
-
1
79
FQ209677.1
-
1
79
FQ209716.1
-
1
79
FQ209779.1
-
1
79
FQ209814.1
-
1
79
FQ209897.1
-
1
79
FQ209914.1
-
1
79
FQ210068.1
-
1
79
FQ210169.1
-
1
79
FQ210655.1
-
2
558
FQ210685.1
-
1
79
FQ210775.1
-
1
79
FQ210989.1
-
1
79
FQ210996.1
-
1
79
FQ211012.1
-
1
79
FQ211042.1
-
1
79
FQ211203.1
-
1
79
FQ211353.1
-
1
79
FQ218241.1
-
1
79
FQ218252.1
-
1
79
FQ218261.1
-
1
79
FQ218411.1
-
2
697
FQ218460.1
-
1
79
FQ218555.1
-
1
79
FQ218567.1
-
1
79
FQ218586.1
-
1
79
FQ218654.1
-
1
79
FQ218694.1
-
1
79
FQ218710.1
-
1
79
FQ218855.1
-
1
79
FQ218857.1
-
1
79
FQ218947.1
-
1
79
FQ218955.1
-
1
79
FQ219224.1
-
1
79
FQ219252.1
-
1
79
FQ219268.1
-
1
79
FQ219276.1
-
1
79
FQ219303.1
-
1
79
FQ219383.1
-
1
79
FQ219441.1
-
1
79
FQ219458.1
-
1
79
FQ219621.1
-
1
79
FQ219652.1
-
1
79
FQ219732.1
-
1
79
FQ219741.1
-
2
559
FQ219765.1
-
1
79
M32247.1
-
1
505
Back To Top
More Information
Panther Classification:
Molecular Function -
protease inhibitor
Gene Ontology Categories:
Function(s)
Process(es)
response to hypoxia
acute-phase response
inflammatory response
response to inorganic substance
response to lead ion
response to organic cyclic compound
response to estradiol
response to lipopolysaccharide
response to methanol
response to triglyceride
response to cytokine
response to chromate
negative regulation of serine-type endopeptidase activity
glycoprotein binding
protease binding
endopeptidase inhibitor activity
serine-type endopeptidase inhibitor activity
protein binding
identical protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0