Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNTRL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007018.4 | NP_008949.4 | 30, 31 | 5112 | ||
XM_005251679.3 | XP_005251736.1 | 32, 33 | 5384 | |||
XM_006716940.2 | XP_006717003.1 | 32, 33 | 5384 | |||
XM_006716942.1 | XP_006717005.1 | 21, 22 | 3613 | |||
XM_011518166.1 | XP_011516468.1 | 32, 33 | 5384 | |||
XM_011518167.1 | XP_011516469.1 | 31, 32 | 5225 | |||
XM_011518168.1 | XP_011516470.1 | 32, 33 | 5309 | |||
XM_011518169.2 | XP_011516471.1 | 31, 32 | 5259 | |||
XM_011518170.1 | XP_011516472.1 | 31, 32 | 5207 | |||
XM_011518173.1 | XP_011516475.1 | 20, 21 | 3459 | |||
XM_017014223.1 | XP_016869712.1 | 32, 33 | 5384 | |||
XM_017014224.1 | XP_016869713.1 | 32, 33 | 5384 | |||
XM_017014225.1 | XP_016869714.1 | 30, 31 | 5070 | |||
XM_017014226.1 | XP_016869715.1 | 21, 22 | 3613 | |||
GenBank mRNA | AF083322.1 | - | 7, 8 | 1560 | ||
AF513978.1 | - | 30, 31 | 5081 | |||
AK074079.1 | - | 21, 22 | 3485 | |||
AK297330.1 | - | 1432 | ||||
BC137284.1 | - | 30, 31 | 5105 | 9021281 | ||
BC137286.1 | - | 30, 31 | 5104 | 9021283 |