Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098783.2 | NP_001092253.1 | 4 | 615 | ||
NM_007042.4 | NP_008973.1 | 4 | 381 | |||
NR_049755.1 | 3 | 517 | ||||
NR_049756.1 | 4 | 381 | ||||
NR_049757.1 | 3 | 283 | ||||
NR_049758.1 | 2 | 198 | ||||
XM_011533313.2 | XP_011531615.1 | 4 | 415 | |||
XM_017005641.1 | XP_016861130.1 | 4 | 378 | |||
GenBank mRNA | AF001175.1 | - | 4 | 356 | ||
AK311396.1 | - | 3 | 242 | |||
AK311969.1 | - | 4 | 310 | |||
AV760694.2 | - | 2 | 181 | |||
BC002441.1 | - | 4 | 329 | 3347003 | ||
BC007342.1 | - | 4 | 329 | 3616325 | ||
BC012017.1 | - | 4 | 342 | 4708724 | ||
BP352444.1 | - | 3 | 266 | |||
CD698741.1 | - | 4 | 371 | |||
CN354601.1 | - | 4 | 308 | |||
CR407611.1 | - | 3 | 187 | |||
DA040251.1 | - | 4 | 345 | |||
DC303206.1 | - | 3 | 452 | |||
DC360931.1 | - | 3 | 517 |