Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000065.3 | NP_000056.2 | 8 | 1223 | ||
NM_001115131.2 | NP_001108603.2 | 8 | 1250 | |||
XM_005248357.2 | XP_005248414.1 | 8 | 1298 | |||
XM_006714496.3 | XP_006714559.1 | 8 | 1298 | |||
XM_011514114.2 | XP_011512416.1 | 8 | 1298 | |||
XM_011514115.2 | XP_011512417.1 | 8 | 1219 | |||
XM_011514116.2 | XP_011512418.1 | 9 | 1335 | |||
XM_011514117.2 | XP_011512419.1 | 9 | 1377 | |||
XM_011514118.2 | XP_011512420.1 | 8 | 1115 | |||
XM_011514119.2 | XP_011512421.1 | 8 | 1298 | |||
XM_011514121.2 | XP_011512423.1 | 3 | 384 | |||
XM_017009818.1 | XP_016865307.1 | 9 | 1358 | |||
XM_017009819.1 | XP_016865308.1 | 4 | 561 | |||
GenBank mRNA | AB126592.1 | - | 7 | 985 | ||
AK292509.1 | - | 8 | 1121 | |||
AK313673.1 | - | 8 | 1142 | |||
BC035723.1 | - | 8 | 1250 | 5591446 | ||
J04506.1 | - | 8 | 1051 | |||
J05024.1 | - | 8 | 1042 | |||
J05064.1 | - | 8 | 1140 |