Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000562.2 | NP_000553.1 | 8 | 1249 | ||
GenBank mRNA | BC020702.1 | - | 8 | 1222 | ||
BC132911.1 | - | 8 | 1141 | 40146933 | ||
BC132913.1 | - | 8 | 1141 | 40146935 | ||
M16974.1 | - | 8 | 1254 |