Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC7A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126335.1 | NP_001119807.1 | 10, 11 | 1201 | ||
NM_001243036.1 | NP_001229965.1 | 10, 11 | 1296 | |||
NM_014270.4 | NP_055085.1 | 10, 11 | 1287 | |||
XM_006722992.1 | XP_006723055.1 | 8, 9 | 962 | |||
XM_011526402.2 | XP_011524704.1 | 10, 11 | 2391 | |||
XM_017026230.1 | XP_016881719.1 | 8, 9 | 937 | |||
GenBank mRNA | AB033548.1 | - | 10, 11 | 1172 | ||
AF141289.1 | - | 10, 11 | 1269 | |||
AJ249199.1 | - | 9, 10 | 1146 | |||
AK026446.1 | - | 10, 11 | 1270 | |||
AK223147.1 | - | 10, 11 | 1275 | |||
AK313708.1 | - | 10, 11 | 1188 | |||
AL365340.1 | - | 3, 4 | 339 | |||
AL365341.1 | - | 2, 3 | 123 | |||
BC017962.1 | - | 10, 11 | 1276 | 4608486 |