Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nkx2-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013093.1 | NP_037225.1 | 2 | 2035 | ||
XM_006240116.2 | XP_006240178.1 | 3 | 2316 | |||
XM_006240117.2 | XP_006240179.1 | 3 | 2389 | |||
XM_006240118.2 | XP_006240180.1 | 3 | 2900 | |||
GenBank mRNA | AB894119.1 | - | 2 | 2034 | ||
X53858.1 | - | 2 | 2097 |