Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC498465 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC079195.1 | - | 5 | 1057 |
Gene Symbol: LOC102546376 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770586.1 | XP_008768808.1 | 5 | 1291 |
Gene Symbol: LOC102546495 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006251679.2 | XP_006251741.1 | 4 | 666 | ||
XM_008770588.1 | XP_008768810.1 | 5 | 891 |
Gene Symbol: LOC102550466 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770593.1 | XP_008768815.1 | 5 | 1196 |