Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTP4A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007079.3 | NP_009010.2 | 1, 2 | 956 | ||
NM_032611.2 | NP_116000.1 | 1, 2 | 956 | |||
XM_005250764.2 | XP_005250821.1 | 3, 4 | 1341 | |||
XM_011516803.1 | XP_011515105.1 | 2, 3 | 1218 | |||
XM_011516805.1 | XP_011515107.1 | 3, 4 | 475 | |||
XM_017012998.1 | XP_016868487.1 | 2, 3 | 1048 | |||
GenBank mRNA | AF041434.1 | - | 1, 2 | 340 | ||
AJ276554.1 | - | 1, 2 | 326 | |||
BC003105.1 | - | 1, 2 | 437 | 3357244 | ||
BT007303.1 | - | 1, 2 | 103 |