Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RABL2B SIRNA Assays ›
Gene Symbol:
RABL2B
RABL2A
Gene Name:
RAB, member of RAS oncogene family-like 2B
RAB, member of RAS oncogene family-like 2A
Gene Aliases:
-
-
Chromosome Location:
Chr.22: 50767492 - 50783705 on Build GRCh38
Chromosome Location:
Chr.2: 113627218 - 113643398 on Build GRCh38
UniGene ID:
Hs.584862
Hs.446425
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID
s22013
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RABL2B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001003789.1
NP_001003789.1
9
1416
NM_001130919.1
NP_001124391.1
9
1412
NM_001130920.1
NP_001124392.1
10
1571
NM_001130921.1
NP_001124393.1
10
1664
NM_001130922.1
NP_001124394.1
9
1413
NM_001130923.1
NP_001124395.1
10
1598
NM_007081.2
NP_009012.1
9
1409
XM_005261899.2
XP_005261956.1
10
1695
XM_011530668.1
XP_011528970.1
10
1698
XM_011530669.2
XP_011528971.1
9
1445
XM_011530670.1
XP_011528972.1
9
1462
XM_011530671.1
XP_011528973.1
10
1572
XM_011530673.1
XP_011528975.1
10
1686
XM_011530675.1
XP_011528977.1
10
1665
XM_017028543.1
XP_016884032.1
10
1888
XM_017028544.1
XP_016884033.1
10
1793
XM_017028545.1
XP_016884034.1
10
1734
XM_017028546.1
XP_016884035.1
10
1736
XM_017028547.1
XP_016884036.1
10
1885
XM_017028548.1
XP_016884037.1
9
1442
XM_017028549.1
XP_016884038.1
10
1731
XM_017028550.1
XP_016884039.1
10
1560
XM_017028551.1
XP_016884040.1
8
1604
XM_017028552.1
XP_016884041.1
10
1858
XM_017028553.1
XP_016884042.1
10
1760
XM_017028554.1
XP_016884043.1
10
1704
XM_017028555.1
XP_016884044.1
10
1706
XM_017028556.1
XP_016884045.1
10
1855
XM_017028557.1
XP_016884046.1
10
1701
XM_017028558.1
XP_016884047.1
10
1539
GenBank mRNA
AF095352.1
-
9
1356
AK094341.1
-
3
1738
BC014879.2
-
9
1377
3906749
BC024281.1
-
10
1614
4580599
Gene Symbol: RABL2A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001306158.1
NP_001293087.1
9
1418
NM_001306161.1
NP_001293090.1
7
1230
NM_013412.3
NP_038198.1
9
1415
XM_005263569.3
XP_005263626.1
9
1459
XM_005263570.3
XP_005263627.1
9
1426
XM_006712207.3
XP_006712270.1
9
1528
XM_006712208.3
XP_006712271.1
9
1417
XM_006712209.3
XP_006712272.1
9
1571
XM_006712215.3
XP_006712278.1
10
1628
XM_011510509.2
XP_011508811.1
9
1419
XM_017003203.1
XP_016858692.1
8
2168
XM_017003204.1
XP_016858693.1
9
2108
XM_017003206.1
XP_016858695.1
10
1569
XM_017003207.1
XP_016858696.1
9
1482
XM_017003208.1
XP_016858697.1
9
1439
XM_017003209.1
XP_016858698.1
9
1423
XM_017003210.1
XP_016858699.1
8
2156
XM_017003211.1
XP_016858700.1
9
1447
XM_017003212.1
XP_016858701.1
8
2153
XM_017003213.1
XP_016858702.1
8
2138
XM_017003214.1
XP_016858703.1
9
2078
XM_017003216.1
XP_016858705.1
9
1541
XM_017003217.1
XP_016858706.1
9
1425
XM_017003218.1
XP_016858707.1
10
1579
XM_017003219.1
XP_016858708.1
9
1492
XM_017003220.1
XP_016858709.1
8
2135
XM_017003221.1
XP_016858710.1
9
1411
XM_017003222.1
XP_016858711.1
10
1566
XM_017003223.1
XP_016858712.1
6
1976
XM_017003224.1
XP_016858713.1
7
1243
XM_017003225.1
XP_016858714.1
7
1234
XM_017003226.1
XP_016858715.1
6
1946
XM_017003227.1
XP_016858716.1
7
1213
XM_017003228.1
XP_016858717.1
8
1341
XM_017003229.1
XP_016858718.1
9
1435
XM_017003230.1
XP_016858719.1
9
1449
XM_017003232.1
XP_016858721.1
8
1296
GenBank mRNA
AF095351.1
-
9
1349
AK092971.1
-
3
2147
AK289496.1
-
10
1333
AL137649.1
-
5
1016
BC111008.1
-
7
1220
5769737
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
small GTPase mediated signal transduction
GTPase activity
GTP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0