Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOX21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007084.3 | NP_009015.1 | 1 | 1919 | ||
GenBank mRNA | BM783159.1 | - | 1 | 383 |