Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ipmk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134417.1 | NP_599244.1 | 6 | 933 | ||
XM_006256345.2 | XP_006256407.1 | 6 | 989 | |||
XM_006256346.2 | XP_006256408.1 | 6 | 1351 | |||
XM_008772858.1 | XP_008771080.1 | 6 | 865 | |||
GenBank mRNA | AY014898.1 | - | 6 | 933 |