Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lgi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145769.3 | NP_665712.1 | 8 | 1260 | ||
GenBank mRNA | AJ487517.1 | - | 8 | 1260 | ||
BC089222.1 | - | 8 | 1367 | 7320899 |