Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAZ2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300905.1 | NP_001287834.1 | 20 | 4169 | ||
NM_013449.3 | NP_038477.2 | 20 | 4160 | |||
XM_011537797.2 | XP_011536099.1 | 20 | 4074 | |||
XM_011537798.2 | XP_011536100.1 | 20 | 4074 | |||
XM_011537799.2 | XP_011536101.1 | 21 | 3984 | |||
XM_011537800.2 | XP_011536102.1 | 20 | 4126 | |||
XM_011537801.2 | XP_011536103.1 | 20 | 4153 | |||
XM_011537803.2 | XP_011536105.1 | 20 | 4169 | |||
XM_011537804.2 | XP_011536106.1 | 22 | 4252 | |||
XM_011537805.2 | XP_011536107.1 | 21 | 4255 | |||
XM_011537806.2 | XP_011536108.1 | 20 | 4033 | |||
XM_011537807.2 | XP_011536109.1 | 19 | 3840 | |||
XM_011537809.2 | XP_011536111.1 | 14 | 2373 | |||
XM_017018740.1 | XP_016874229.1 | 21 | 4300 | |||
XM_017018741.1 | XP_016874230.1 | 21 | 4074 | |||
XM_017018742.1 | XP_016874231.1 | 22 | 4210 | |||
GenBank mRNA | AB002312.2 | - | 19 | 3949 | ||
AK023830.1 | - | 18 | 3795 | |||
AK023842.1 | - | 20 | 4047 | |||
BC008965.2 | - | 4 | 774 | |||
CR749379.1 | - | 20 | 4161 |