Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nkain4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006235744.2 | XP_006235806.1 | 2 | 241 | ||
XM_006235745.2 | XP_006235807.1 | 2 | 2123 | |||
XM_008762496.1 | XP_008760718.1 | 3 | 590 | |||
XM_008762497.1 | XP_008760719.1 | 3 | 1418 | |||
XM_008762498.1 | XP_008760720.1 | 3 | 393 |