Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A1i3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037975.2 | NP_001033064.2 | 16 | 2002 | ||
XM_006237281.2 | XP_006237343.1 | 16 | 2014 | |||
GenBank mRNA | J03552.1 | - | 16 | 2002 |