Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGD1559786 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034132.1 | NP_001029304.1 | 6 | 308 | ||
XM_006238508.2 | XP_006238570.1 | 6 | 424 | |||
XM_006238509.2 | XP_006238571.1 | 5 | 499 | |||
XM_008763867.1 | XP_008762089.1 | 5 | 267 | |||
GenBank mRNA | BC100100.1 | - | 6 | 308 | 7376782 | |
FQ228722.1 | - | 6 | 327 |