Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303029.1 | NP_001289958.1 | 5 | 1114 | ||
NM_007183.3 | NP_009114.1 | 4 | 1150 | |||
GenBank mRNA | AF053719.1 | - | 4 | 1094 | ||
AK025067.1 | - | 4 | 1111 | |||
AK223511.1 | - | 4 | 1111 | |||
BC000081.2 | - | 4 | 1116 | 3504300 | ||
FN421477.1 | - | 5 | 1114 | |||
Z98265.1 | - | 4 | 1114 |