Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbx22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106937.1 | NP_001100407.1 | 5 | 555 | ||
XM_006257159.2 | XP_006257221.1 | 6 | 824 | |||
XM_006257160.2 | XP_006257222.1 | 6 | 674 | |||
XM_006257161.2 | XP_006257223.1 | 6 | 821 | |||
XM_006257162.2 | XP_006257224.1 | 5 | 864 | |||
XM_008773339.1 | XP_008771561.1 | 5 | 539 |