Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282291.1 | NP_001269220.1 | 3 | 493 | ||
NM_001282292.1 | NP_001269221.1 | 2 | 442 | |||
NM_007188.4 | NP_009119.2 | 2 | 442 | |||
GenBank mRNA | AF047690.1 | - | 2 | 392 | ||
AK002018.1 | - | 3 | 465 | |||
AK024401.1 | - | 2 | 362 | |||
AK094636.1 | - | 3 | 370 | |||
AK222911.1 | - | 2 | 414 | |||
AK314721.1 | - | 2 | 414 | |||
BC141814.1 | - | 2 | 402 | 40147981 | ||
BC141836.1 | - | 2 | 402 | 40148945 | ||
BC151235.1 | - | 2 | 404 | 8860430 | ||
DA147595.1 | - | 2 | 441 | |||
HY000407.1 | - | 2 | 442 |
Gene Symbol: ASIC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK128129.1 | - | 3 | 552 |